Cortisol express ELISA kit
Cortisol Express ELISA kit offers a fast, convenient assay format for measurement of cortisol.
- Measure cortisol in urine, plasma, and other sample matrices
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure cortisol levels down to 110 pg/ml
- Rapid assay; get results in under four hours
Cortisol Express ELISA is a competitive assay that permits the rapid measurement of cortisol from biological samples, requiring only a two-hour incubation and one-hour development times. This ELISA offers the convenience of a fast assay while maintaining sensitivity.
: Kit contains monoclonal antibody, AChE tracer, standard, ELISA and wash buffer, polysorbate 20, coated plate, cover sheet, ellman's reagent, tracer and antiserum dye.
: Due to an approximate 15% cross-reactivity with dexamethasone, this kit may not be suitable for use with samples that contain high concentrations of dexamethasone, such as those collected during a dexamethasone suppression test.
- Pakn:480 Tests
- Beskrivelse:Cortisol Express ELISA kit, 480 soild wells
- Cat. No.:CAYM500370-480S
- Mid point:300 - 500 pg/ml (50% B/B₀)
- Sensitivitet:110 pg/ml (80% B/B₀)
- Detektionsområde:39,1 - 5000 pg/ml
- Udviklingstid:120 min
- Opbevaringstemperatur:–20 °C
- Inkubationstid:2 hr



