Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cortisol express ELISA kit

Cortisol express ELISA kit

Cortisol Express ELISA kit, 96 strip wells, 96 Tests
Katalog # CAYM500370-96
Leverandør:   Cayman Chemical
Cortisol express ELISA kit
Katalog # CAYM500370-96
Leverandør:  Cayman Chemical
CAS nummer:  
Cortisol express ELISA kit
Cortisol express ELISA kit

Cortisol Express ELISA kit, 96 strip wells, 96 Tests

CAS nummer:  
Price
3.620,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Cortisol Express ELISA kit offers a fast, convenient assay format for measurement of cortisol.

  • Measure cortisol in urine, plasma, and other sample matrices
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure cortisol levels down to 110 pg/ml
  • Rapid assay; get results in under four hours

Cortisol Express ELISA is a competitive assay that permits the rapid measurement of cortisol from biological samples, requiring only a two-hour incubation and one-hour development times. This ELISA offers the convenience of a fast assay while maintaining sensitivity.

Emballering: Kit contains monoclonal antibody, AChE tracer, standard, ELISA and wash buffer, polysorbate 20, coated plate, cover sheet, ellman's reagent, tracer and antiserum dye.

Advarsel: Due to an approximate 15% cross-reactivity with dexamethasone, this kit may not be suitable for use with samples that contain high concentrations of dexamethasone, such as those collected during a dexamethasone suppression test.

Specifikationer

Specifikationer

  • Pakn:
    96 Tests
  • Beskrivelse:
    Cortisol Express ELISA kit, 96 strip wells
  • Cat. No.:
    CAYM500370-96
  • Mid point:
    300 - 500 pg/ml (50% B/B₀)
  • Sensitivitet:
    110 pg/ml (80% B/B₀)
  • Format:
    Pre-coated
  • Detektionsområde:
    39,1 - 5000 pg/ml
  • Target protein:
    Cortisol
  • Udviklingstid:
    120 min
  • Opbevaringstemperatur:
    –20 °C
  • Analysetype:
    Competitive
  • Inkubationstid:
    2 hr

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.