Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
1-Benzylimidazol

1-Benzylimidazol

Glass bottle for solids, 25 g
Katalog # CAYM70510-25
Leverandør:   Cayman Chemical
1-Benzylimidazol
Katalog # CAYM70510-25
Leverandør:  Cayman Chemical
CAS nummer:  4238-71-5
1-Benzylimidazol

Glass bottle for solids, 25 g

CAS nummer:  4238-71-5
Price
1.280,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  1-Benzyl-1H-imidazole
  • CAS nummer:4238-71-5
  • Formel:C₁₀H₁₀N₂
  • MDL nummer:MFCD00005296
  • Molecular Weight:158.2 g/mol
  • Kogende Pt:310 °C (1013 hPa)
  • Smeltende Pt:72 °C
  • Tæthed:1,22 g/cm³ (25 °C)
  • EINECS:224-200-4
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.