Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Thiol Detection Assay Kits

Thiol Detection Assay Kits

Thiol detection assay kit, 40 samples, 1
Katalog # CAYM700340-480
Leverandør:   Cayman Chemical
Thiol Detection Assay Kits
Katalog # CAYM700340-480
Leverandør:  Cayman Chemical
CAS nummer:  
Thiol Detection Assay Kits
Thiol Detection Assay Kits

Thiol detection assay kit, 40 samples, 1

CAS nummer:  
Price
9.910,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

A simple, reproducible, and sensitive tool for assaying free thiol content in samples.

  • Measure free thiol content in plasma, serum, urine, cell lysates, and tissue homogenates
  • Assay 40 samples in duplicate
  • Plate-based fluorometric measurement (ex 380 to 390 nm, em 510 to 520 nm)
  • Includes both a glutathione and cysteine standard
  • Range: 15 nm to 1 µm

The detection and measurement of free thiols (i.e., free cysteine, glutathione, and cysteine residues on proteins) is one of the essential tasks for investigating biological processes and events in many biological systems. Cayman’s Thiol detection provides a simple, reproducible, and sensitive tool for assaying free thiol content in samples (i.e., plasma, serum, tissue homogenates, cell lysates, and urine). The assay utilises a proprietary fluorometric detector that reacts with thiol groups to emit a strong fluorescent signal which can be detected using excitation wavelengths between 380 to 390 nm and emission wavelengths between 510 to 520 nm.

Delivery information: Kit contains: Buffer, standards, and plate.

Specifikationer

Specifikationer

  • Pakn:
    1
  • Beskrivelse:
    Thiol detection assay kit
  • Test pr. sæt:
    40 samples

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.