Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Buffers, potassium phosphate buffer (0,5 mol/L) solutions

Buffers, potassium phosphate buffer (0,5 mol/L) solutions

Potassium phosphate buffer solution (0,5 mol/l), 9,0, Glass bottle, 500 mL
Katalog # CAYM600210-500
Leverandør:   Cayman Chemical
Buffers, potassium phosphate buffer (0,5 mol/L) solutions
Katalog # CAYM600210-500
Leverandør:  Cayman Chemical
CAS nummer:  
Buffers, potassium phosphate buffer (0,5 mol/L) solutions

Potassium phosphate buffer solution (0,5 mol/l), 9,0, Glass bottle, 500 mL

CAS nummer:  
Price
987,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL
  • Beskrivelse:
    Potassium phosphate buffer solution (0,5 mol/l)
  • pH-værdi:
    9,0

Specifikationer