Buffers, potassium phosphate buffer (0,5 mol/L) solutions
Potassium phosphate buffer solution (0,5 mol/l), 9,0, Glass bottle, 250 ml
Katalog # CAYM600210-250
Leverandør: Cayman Chemical
Buffers, potassium phosphate buffer (0,5 mol/L) solutions
Katalog # CAYM600210-250
Leverandør: Cayman Chemical
CAS nummer:
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:250 ml
- Beskrivelse:Potassium phosphate buffer solution (0,5 mol/l)
- pH-værdi:9,0
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



