Buffers, potassium phosphate buffer (0,5 mol/L) solutions
Potassium phosphate buffer solution (0,5 mol/l), 7,0, Glass bottle, 500 ml
Catalog # CAYM600208-500
Supplier: Cayman Chemical
Buffers, potassium phosphate buffer (0,5 mol/L) solutions
Catalog # CAYM600208-500
Supplier: Cayman Chemical
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pakning:Glass bottle
- Pakn:500 ml
- Beskrivelse:Potassium phosphate buffer solution (0,5 mol/l)
- pH-værdi:7,0



