Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
γ-Linolensyre

γ-Linolensyre

Glass bottle for solids, 1 g
Catalog # CAYM90220-1
Supplier:   Cayman Chemical
γ-Linolensyre
Catalog # CAYM90220-1
Supplier:  Cayman Chemical
CAS Number:  506-26-3
γ-Linolensyre

Glass bottle for solids, 1 g

CAS Number:  506-26-3
Price
2.120,00 kr
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  cis,cis,cis-6,9,12-Octadecatrienoic acid, γ-Linolenic acid, (6Z,9Z,12Z)-Octadeca-6,9,12-trienoic acid, 6Z,9Z,12Z-Octadecatrienoic acid
  • CAS Number:506-26-3
  • Formula:C₁₈H₃₀O₂
  • MDL Number:MFCD00065718
  • Molecular Weight:278.44 g/mol
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifications