7-Nitroindazol
Glass bottle for solids, 250 mg
Katalog # CAYM81340-250
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
Synonymer: 7-Nitroindazole, 7-NINA, 7-Nitro-1H-indazole
- CAS nummer:2942-42-9
- Formel:C₇H₅N₃O₂
- MDL nummer:MFCD00022789
- Molecular Weight:163.14 g/mol
- Smeltende Pt:190 °C
- Tæthed:1,56 g/cm³ (293 °C)
- EINECS:220-934-4
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:250 mg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



