Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

PPM-18 ≥95%

Glass vial, 10 mg
Katalog # CAYM13327-10
Leverandør:   Cayman Chemical
PPM-18 ≥95%
Katalog # CAYM13327-10
Leverandør:  Cayman Chemical
CAS nummer:  65240-86-0
PPM-18 ≥95%

Glass vial, 10 mg

CAS nummer:  65240-86-0
Price
1.680,00 kr
Log ind for tilgængelighed
1 * 10 mg
Product Details & Documents
  • Inhibits activation of NF-κB

An inhibitor NF-κB activation in vitro and in vivo (IC₅₀= 5 µm).

Synonymer:  NSC 73233
  • CAS nummer:65240-86-0
  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationstestresultater

  • Formulation:
    A crystalline solid
  • Purity:
    ≥95%
  • Stability:
    ≥2 years

Specifikationer

  • Pakning:
    Glass vial
  • Pakn:
    10 mg

Specifikationer