Arecoliniumbromid
Glass bottle for solids, 100 mg
Katalog # CAYM13662-100
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
Synonymer: Arecoline hydrobromide
- CAS nummer:300-08-3
- Formel:C₈H₁₄BrNO₂
- MDL nummer:MFCD00039041
- Molecular Weight:236.11 g/mol
- EINECS:206-087-3
- FN:1544
- ADR:6.1,III
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 mg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



