Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Lansoprazole

Lansoprazole

Glass bottle for solids, 10 g
Katalog # CAYM13627-10
Leverandør:   Cayman Chemical
Lansoprazole
Katalog # CAYM13627-10
Leverandør:  Cayman Chemical
CAS nummer:  103577-45-3
Lansoprazole

Glass bottle for solids, 10 g

CAS nummer:  103577-45-3
Price
5.930,00 kr
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents
Synonymer:  2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole
  • CAS nummer:103577-45-3
  • Formel:C₁₆H₁₄F₃N₃O₂S
  • MDL nummer:MFCD00866873
  • Molecular Weight:369.36 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.