Lansoprazole
Glass bottle for solids, 5 g
Katalog # CAYM13627-5
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
Synonymer: 2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole
- CAS nummer:103577-45-3
- Formel:C₁₆H₁₄F₃N₃O₂S
- MDL nummer:MFCD00866873
- Molecular Weight:369.36 g/mol
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 g



