Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Lansoprazole

Lansoprazole

Glass bottle for solids, 1 g
Catalog # CAYM13627-1
Supplier:   Cayman Chemical
Lansoprazole
Catalog # CAYM13627-1
Supplier:  Cayman Chemical
CAS Number:  103577-45-3
Lansoprazole

Glass bottle for solids, 1 g

CAS Number:  103577-45-3
Price
850,00 kr
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole
  • CAS Number:103577-45-3
  • Formula:C₁₆H₁₄F₃N₃O₂S
  • MDL Number:MFCD00866873
  • Molecular Weight:369.36 g/mol
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.