Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Tetranor PGDM ≥90%

Tetranor PGDM ≥90%

25 µG
Katalog # CAYM12850-25
Leverandør:   Cayman Chemical
Tetranor PGDM ≥90%
Katalog # CAYM12850-25
Leverandør:  Cayman Chemical
CAS nummer:  70803-91-7
Tetranor PGDM ≥90%

25 µG

CAS nummer:  70803-91-7
Price
1.970,00 kr
Log ind for tilgængelighed
1 * 25 µl
Product Details & Documents

Tetranor PGDM is a major metabolite of prostaglandin D2 which are mainly involved in the inhibition of platelet aggregation and relaxation of vascular smooth muscle.

  • Formulated as a solution in methanol

The levels of tetranor PGDM is found to be as 1,5±0,3 ng/mg creatinine and 8,1±1,3 ng/mg creatinine in human and mouse urine respectively.

Synonymer:  tetranor-Prosatglandin D Metabolite, tetranor-PGD Metabolite
  • CAS nummer:70803-91-7
  • Formel:C₁₆H₂₄O₇
  • Molecular Weight:328.4 g/mol
  • Opbevaringstemperatur:Ultra-lav temperatur fryser (–80°C)

Documents

Specifikationer

Specifikationer

  • Pakn:
    25 µG

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.