Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(±)-20-HDoHE

(±)-20-HDoHE

100 µG
Catalog # CAYM33750-100
Supplier:   Cayman Chemical
(±)-20-HDoHE
Catalog # CAYM33750-100
Supplier:  Cayman Chemical
CAS Number:  90906-41-5
(±)-20-HDoHE

100 µG

CAS Number:  90906-41-5
Price
3.310,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
  • CAS Number:90906-41-5
  • Formula:C₂₂H₃₂O₃
Specifications

Specifications

  • Pakn:
    100 µG

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.