Orders LinkContactUsLinkComponent
(±)-17-HDoHE
(±)-17-HDoHE
100 µG
Catalog # CAYM33650-100
Supplier:   Cayman Chemical
(±)-17-HDoHE
Catalog # CAYM33650-100
Supplier:  Cayman Chemical
CAS Number:  90780-52-2
(±)-17-HDoHE
100 µG
CAS Number:  90780-52-2
Price
3.270,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
  • CAS Number:90780-52-2
  • Formula:C₂₂H₃₂O₃
Specifications
Specifications
  • Pakn:
    100 µG
  • Cat. No.:
    CAYM33650-100
Specifications