Hexacosansyre
Glass bottle for solids, 250 mg
Catalog # CAYM13354-250
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: Cerotic acid, Hexacosanoic acid
- CAS Number:506-46-7
- Formula:C₂₆H₅₂O₂
- MDL Number:MFCD00002811
- Molecular Weight:396.7 g/mol
- Boiling Pt:419 °C (1013 hPa)
- Melting Pt:76…77 °C
- Density:0,8198 g/cm³ (100 °C)
- Storage Temperature:Køleskab
- EINECS:208-040-2
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:250 mg



