Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(±)-4-HDoHE

(±)-4-HDoHE

100 µG
Katalog # CAYM33200-100
Leverandør:   Cayman Chemical
(±)-4-HDoHE
Katalog # CAYM33200-100
Leverandør:  Cayman Chemical
CAS nummer:  90906-40-4
(±)-4-HDoHE

100 µG

CAS nummer:  90906-40-4
Price
3.310,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
  • CAS nummer:90906-40-4
  • Formel:C₂₂H₃₂O₃
Specifikationer

Specifikationer

  • Pakn:
    100 µG

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.