Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(±)-4-HDoHE

(±)-4-HDoHE

50 µG
Katalog # CAYM33200-50
Leverandør:   Cayman Chemical
(±)-4-HDoHE
Katalog # CAYM33200-50
Leverandør:  Cayman Chemical
CAS nummer:  90906-40-4
(±)-4-HDoHE

50 µG

CAS nummer:  90906-40-4
Price
1.760,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
  • CAS nummer:90906-40-4
  • Formel:C₂₂H₃₂O₃
Specifikationer

Specifikationer

  • Pakn:
    50 µG

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.