Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
GN25

GN25

50 mg
Katalog # CAYM10948-50
Leverandør:   Cayman Chemical
GN25
Katalog # CAYM10948-50
Leverandør:  Cayman Chemical
CAS nummer:  1227401-27-5
CAS nummer:  1227401-27-5
Price
3.200,00 kr
Log ind for tilgængelighed
1 * 50 mg
Product Details & Documents

A 2-thio-dimethoxy naphthoquinone analog that blocks Snail binding to p53 and induces p53 expression in cancer cells in a K-Ras dependent manner

Synonymer:  3-​[(1,​4-​dihydro-​5,​8-​dimethoxy-​1,​4-​dioxo-​2-​naphthalenyl)thio]-​propanoic acid, p53-Snail binding Inhibitor
  • CAS nummer:1227401-27-5
  • Formel:C₁₅H₁₄O₆S
  • Molecular Weight:322.3 g/mol
Specifikationer

Specifikationer

  • Pakn:
    50 mg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.