GN25
10 mg
Katalog # CAYM10948-10
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
A 2-thio-dimethoxy naphthoquinone analog that blocks Snail binding to p53 and induces p53 expression in cancer cells in a K-Ras dependent manner
Synonymer: 3-[(1,4-dihydro-5,8-dimethoxy-1,4-dioxo-2-naphthalenyl)thio]-propanoic acid, p53-Snail binding Inhibitor
- CAS nummer:1227401-27-5
- Formel:C₁₅H₁₄O₆S
- Molecular Weight:322.3 g/mol
Specifikationer
Specifikationer
- Pakn:10 mg



