Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
(10S,17S)-DiHDoHE

(10S,17S)-DiHDoHE

100 µG
Catalog # CAYM10008128-100
Supplier:   Cayman Chemical
(10S,17S)-DiHDoHE
Catalog # CAYM10008128-100
Supplier:  Cayman Chemical
CAS Number:  871826-47-0
(10S,17S)-DiHDoHE

100 µG

CAS Number:  871826-47-0
Price
7.390,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
  • CAS Number:871826-47-0
  • Formula:C₂₂H₃₂O₄
Specifications

Specifications

  • Pakn:
    100 µG

Specifications