Fenofibrate
50 g
CAYM10005368-50
2-[4-(4-chlorobenzoyl)phenoxy]-2-methyl-propanoic acid, 1-methylethyl ester
: 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid Isopropyl Ester, Isopropyl 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate, Isopropyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionate
- :49562-28-9
- :C₂₀H₂₁ClO₄
- :MFCD00133314
- :360.83 g/mol
- Formula weight:360.8
- Purity:≥98%
- Formulation:Crystalline solid
- Storage temperature:-20°C
- Stability:≥ 2 years
- λmax:287 nm
- Pakn:50 g



