Fenofibrate
1 g
Katalog # CAYM10005368-1
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
2-[4-(4-chlorobenzoyl)phenoxy]-2-methyl-propanoic acid, 1-methylethyl ester
Synonymer: 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic Acid Isopropyl Ester, Isopropyl 2-(4-(4-chlorobenzoyl)phenoxy)-2-methylpropanoate, Isopropyl 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionate
- CAS nummer:49562-28-9
- Formel:C₂₀H₂₁ClO₄
- MDL nummer:MFCD00133314
- Molecular Weight:360.83 g/mol
Specifikationer
Specifikationstestresultater
- Formula weight:360.8
- Purity:≥98%
- Formulation:Crystalline solid
- Storage temperature:-20°C
- Stability:≥ 2 years
- λmax:287 nm
Specifikationer
- Pakn:1 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



