Orders LinkContactUsLinkComponent
Stearinsyre
Stearinsyre
Glass bottle for solids, 10 g
Catalog # CAYM10011298-10
Supplier:   Cayman Chemical
Stearinsyre
Catalog # CAYM10011298-10
Supplier:  Cayman Chemical
CAS Number:  57-11-4
Stearinsyre
Stearinsyre
Glass bottle for solids, 10 g
CAS Number:  57-11-4
Price
1.070,00 kr
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Octadecanoic acid, Octadecylic acid
  • CAS Number:57-11-4
  • Formula:C₁₈H₃₆O₂
  • MDL Number:MFCD00002752
  • Molecular Weight:284.48 g/mol
  • Merck Index:13,08882
  • Boiling Pt:361 °C (1013 hPa)
  • Melting Pt:67 °C
  • Density:0,94 g/cm³ (20 °C)
  • Flash Pt:196 °C (closed cup)
  • Storage Temperature:Køleskab
  • EINECS:200-313-4
Specifications
Specifications
  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g
Specifications