Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

2-Aminopyrazin

Glass bottle for solids, 25 g
Katalog # CAYM10007093-25
Leverandør:   Cayman Chemical
2-Aminopyrazin
Katalog # CAYM10007093-25
Leverandør:  Cayman Chemical
CAS nummer:  5049-61-6
2-Aminopyrazin

Glass bottle for solids, 25 g

CAS nummer:  5049-61-6
Price
944,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  Aminopyrazine, Pyrazin-2-amine
  • CAS nummer:5049-61-6
  • Formel:C₄H₅N₃
  • MDL nummer:MFCD00006137
  • Molecular Weight:95.1 g/mol
  • Kogende Pt:203 °C (1013 hPa)
  • Smeltende Pt:117…120 °C
  • EINECS:225-748-7
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.