8-Bromisoquinolin
Glass bottle for solids, 500 mg
Katalog # CAYM10007082-500
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
- CAS nummer:63927-22-0
- Formel:C₉H₆BrN
- MDL nummer:MFCD04973298
- Molecular Weight:208.0600 g/mol
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 mg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



