5-Bromisoquinolin
Glass bottle for solids, 500 mg
Catalog # CAYM10007079-500
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
- CAS Number:34784-04-8
- Formula:C₉H₆BrN
- MDL Number:MFCD01646405
- Molecular Weight:208.06 g/mol
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:500 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



