Orders LinkContactUsLinkComponent
Accessories for DOSE IT
Accessories for DOSE IT
Silicone tubing, autoclavable, length: 25 m (bulk roll), 1 mm
Katalog # 710-1029
Leverandør:   INTEGRA Biosciences
Accessories for DOSE IT
Katalog # 710-1029
Leverandør:  INTEGRA Biosciences
Accessories for DOSE IT
Accessories for DOSE IT
Silicone tubing, autoclavable, length: 25 m (bulk roll), 1 mm
Price
1.510,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    Silicone tubing, autoclavable, length: 25 m (bulk roll)
  • Ø indv.:
    1 mm