Orders LinkContactUsLinkComponent
Accessories for DOSE IT
Aspiration, dispensing tube, stainless steel, one end dented, length: 10 cm, 4 mm
  710-1035
Accessories for DOSE IT
  710-1035
Accessories for DOSE IT
Accessories for DOSE IT
Aspiration, dispensing tube, stainless steel, one end dented, length: 10 cm, 4 mm
256,00 kr
1 * 1 Items
Documents
  • Beskrivelse:
    Aspiration, dispensing tube, stainless steel, one end dented, length: 10 cm
  • Ø indv.:
    4 mm