Accessories for DOSE IT
Aspiration, dispensing tube, stainless steel, one end dented, length: 10 cm, 8 mm
Katalog # 710-1037
Leverandør: INTEGRA Biosciences
Gå til:
Product Details & Documents
Documents
Dose It
Videoer
Specifikationer
- Beskrivelse:Aspiration, dispensing tube, stainless steel, one end dented, length: 10 cm
- Ø indv.:8 mm