Orders LinkContactUsLinkComponent
Milk butyrometer
Butyrometer for milk, 0 - 6% : 0,1
  614-0090
Milk butyrometer
  614-0090
Milk butyrometer
Milk butyrometer
Butyrometer for milk, 0 - 6% : 0,1
284,00 kr
1 * 10 Items
Borosilicate glass.
  • With or without stopper
Available with a declaration of conformity per batch and type (in German only).
  • Beskrivelse:
    Butyrometer for milk, 0 - 6% : 0,1
  • Seq:
    04
Accessories
  •  
    Beskrivelse
  • 614-2028
    Stopper, rubber, conical, for all measuring method butyrometers
    1 * 100 Items - 360,00 kr