Orders LinkContactUsLinkComponent
Cream butyrometer, without stopper
Cream butyrometer, without stopper
Butyrometer for cream, Roeder´s, 0 - 5 - 40 % : 0,5
Katalog # 614-0107
Leverandør:   GERBER FUNKE
Cream butyrometer, without stopper
Katalog # 614-0107
Leverandør:  GERBER FUNKE
Cream butyrometer, without stopper
Cream butyrometer, without stopper
Butyrometer for cream, Roeder´s, 0 - 5 - 40 % : 0,5
Price
1.080,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
According to Roeder´s weighing method.
Specifikationer
  • Beskrivelse:
    Butyrometer for cream, Roeder´s, 0 - 5 - 40 % : 0,5
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Weighing boat for butter for butyrometers acc. to Roeder's method
    Weighing boat for butter for butyrometers acc. to Roeder's method
    614-2034
    Log ind for tilgængelighed
    1 * 1 Items - 47,80 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Cream beaker, unperforated, for ice cream and condensed milk butyrometer and cream butyrometer acc. to Roeder´s method
    Cream beaker, unperforated, for ice cream and condensed milk butyrometer and cream butyrometer acc. to Roeder´s method
    614-2032
    Log ind for tilgængelighed
    1 * 1 Items - 18,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Stopper, rubber, with hole, for all weighing method butyrometers
    Stopper, rubber, with hole, for all weighing method butyrometers
    614-2030
    Log ind for tilgængelighed
    1 * 100 Items - 540,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Stopper, rubber, for sealing the bulbs of all weighing method butyrometers
    Stopper, rubber, for sealing the bulbs of all weighing method butyrometers
    614-2029
    Log ind for tilgængelighed
    1 * 100 Items - 180,00 kr