Orders LinkContactUsLinkComponent
Cheese trier
Cheese trier
Cheese trier, 190 mm, 125 mm, 13 mm
Katalog # 300-0143
Leverandør:   GERBER FUNKE
Cheese trier
Katalog # 300-0143
Leverandør:  GERBER FUNKE
Cheese trier
Cheese trier
Cheese trier, 190 mm, 125 mm, 13 mm
Price
1.790,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Chrome-nickel steel. Developed for sampling small cores and testing the readiness of cheeses without cutting them open. Ideal for use in milk-processing industries and cheese dairies.
  • Black plastic handle
  • Two different sizes available
Specifikationer
  • Ø:
    13 mm
  • Neds.- dybde:
    125 mm
  • Beskrivelse:
    Cheese trier
  • Længde:
    190 mm
  • Seq:
    02