Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Spermidin ≥99%

Spermidin ≥99%

Glass bottle for solids, 100 g
Katalog # A19096.22
Ætsende
Leverandør:   Thermo Fisher Scientific
Spermidin ≥99%
Katalog # A19096.22
CAS nummer:  124-20-9
Spermidin ≥99%
Spermidin ≥99%

Glass bottle for solids, 100 g

CAS nummer:  124-20-9
Price
13.380,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 124-20-9 Molecular Formula: H2N(CH2)3NH(CH2)4NH2 Formula Weight: 145.25 Flash Point: 110°(230°F) Refractive Index: 1.4790 Melting Point: 22-25° Boiling Point: 128-130°/14mm Beilstein Reference: 1698591 Merck Reference: 14,8742 UN No.: 2735 MDL No.: MFCD00008229

Synonymer:  N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
  • CAS nummer:124-20-9
  • Formel:C₇H₁₉N₃
  • MDL nummer:MFCD00008229
  • Molecular Weight:145.25 g/mol
  • Merck Index:14,8742
  • Kogende Pt:128…130 °C (14 mmHg)
  • Smeltende Pt:22…25 °C
  • Tæthed:0,925
  • Flash Pt:>110 °C (230 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-689-0
  • ADR:8,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.