Orders LinkContactUsLinkComponent
Urinanalysis Kit, KOVA®
Urinanalysis Kit, KOVA®
Cap, flexible with flip tabs, 500
Catalog # 720-2419
Supplier:   Kova International
Urinanalysis Kit, KOVA®
Catalog # 720-2419
Supplier:  Kova International
CAS Number:  
Urinanalysis Kit, KOVA®
Urinanalysis Kit, KOVA®
Cap, flexible with flip tabs, 500
CAS Number:  
Price
638,00 kr
Log in for availability
1 * 500 Items
Product Details & Documents
Complete urinalysis kit for up to 1000 determinations.
  • 12 ml graduated non-sterile disposable centrifuge tubes manufactured of high-quality, break-resistant clear plastic and marked in 1 ml increments
  • Disposable plastic transfer pipettes that feature a unique lock for one-step, contamination-free decanting, retaining 1 ml of urine after centrifugation
  • Precision manufactured, 10-chambered disposable microscope slides made of optically clear plastic, the Glasstic® slide grid is in the focal plane of the specimen, allowing for use over a range of magnifications
 : Contains 100 slides (10 chambered), 1000 KOVA Petters (pipettes with bulge) and 1000 KOVA tubes.
Specifications
Specifications
  • Pakn:
    500
  • Beskrivelse:
    Cap, flexible with flip tabs