Fernbach culture flasks
2800 ml, with GL45 screw cap, PP
Katalog # 391-0534
Leverandør: SciLabware
Gå til:
Product Details & Documents
Culture flask, made of PYREX® borosilicate glass. For use in preparation of cultures.
- Exhibit 3 baffle indents to provide optimal and consistent agitation
- Large surface area to volume ratio
- Available in two versions, with Delong neck or screw cap
Specifikationer
- Model:with GL45 screw cap, PP
- Kapacitet:2800 ml
- Seq:02