Orders LinkContactUsLinkComponent
Fernbach culture flasks
2800 ml, with GL45 screw cap, PP
Katalog # 391-0534
Leverandør:   SciLabware
Fernbach culture flasks
Katalog # 391-0534
Leverandør:  SciLabware
Fernbach culture flasks
Fernbach culture flasks
2800 ml, with GL45 screw cap, PP
Price
5.230,00 kr
Log ind for tilgængelighed
1 * 2 Items
Product Details & Documents
Culture flask, made of PYREX® borosilicate glass. For use in preparation of cultures.
  • Exhibit 3 baffle indents to provide optimal and consistent agitation
  • Large surface area to volume ratio
  • Available in two versions, with Delong neck or screw cap
Specifikationer
  • Model:
    with GL45 screw cap, PP
  • Kapacitet:
    2800 ml
  • Seq:
    02