Fernbach culture flasks
2800 ml, with Delong neck
391-0533
Culture flask, made of PYREX® borosilicate glass. For use in preparation of cultures.
- Exhibit 3 baffle indents to provide optimal and consistent agitation
- Large surface area to volume ratio
- Available in two versions, with Delong neck or screw cap
- Model:with Delong neck
- Kapacitet:2800 ml