VWR® Basic, Frocks with Velcro®
hvid, XL
113-1991
VWR® Basic SPP frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.
- Snap collar, raglan sleeves, VELCRO® closure, knitted cuffs, no pockets
- Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility that is lot controlled and validated through independent laboratory testing.
Colour: White
- Tøj størrelse:XL
- Farve:hvid
- Køn:Unisex
- Cat. No.:113-1991
- Vægt:48 g/m²
- Outfit applikationer:Cleanroom|Laboratory
- Lukkestørrelse:Med Velcro®
- Lommer:uden lommer