Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Natriumcarbonat

Glass bottle for solids, 500 g
Katalog # RC-125
Irriterende
Leverandør:   G-Biosciences
Natriumcarbonat
Katalog # RC-125
Leverandør:  G-Biosciences
CAS nummer:  497-19-8
Natriumcarbonat
Natriumcarbonat

Glass bottle for solids, 500 g

CAS nummer:  497-19-8
Price
575,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  Calcined soda, Carbonic acid disodium salt
  • CAS nummer:497-19-8
  • Formel:Na₂CO₃
  • MDL nummer:MFCD00003494
  • Molecular Weight:105.99 g/mol
  • Merck Index:14,08596
  • Kogende Pt:1600 °C (1013 hPa)
  • Smeltende Pt:854 °C
  • Tæthed:2,53 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:207-838-8
  • NÅ:01-2119485498-19
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.