D(+)-Sucrose
Glass bottle for solids, 500 g
Katalog # RC-099
Leverandør: G-Biosciences
Gå til:
Product Details & Documents
Synonymer: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
- CAS nummer:57-50-1
- Formel:C₁₂H₂₂O₁₁
- MDL nummer:MFCD00006626
- Molecular Weight:342.3 g/mol
- Merck Index:13,08966
- Smeltende Pt:169…170 °C
- Tæthed:1,29 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-334-9
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



