Orders LinkContactUsLinkComponent
Disposable spin columns for protein purification
Disposable spin columns for protein purification
Spinkolonne, <0,1 ml, samlet volumen på 600 μl, resinvolumen 5 - 100 μl, passer til 1,5 og 2 ml centrifugerør
  786-718
Disposable spin columns for protein purification
  786-718
Disposable spin columns for protein purification
Disposable spin columns for protein purification
Spinkolonne, <0,1 ml, samlet volumen på 600 μl, resinvolumen 5 - 100 μl, passer til 1,5 og 2 ml centrifugerør
421,00 kr
1 * 25 Items
These disposable spin columns are designed for protein purification. These spin columns are uniquely designed, with a snap off end converting to a closure for the column for easy manipulation and use.
These columns can be spun with the stopper attached and then snapped off to elute solutions. For further storage or incubation, simply push the column into the stopper.
  • Beskrivelse:
    Spinkolonne, <0,1 ml, samlet volumen på 600 μl, resinvolumen 5 - 100 μl, passer til 1,5 og 2 ml centrifugerør
  • Seq:
    01