Orders LinkContactUsLinkComponent
Jacalin for IgA purification
Jacalin for IgA purification
Jacalin, immobilised, lyophilised, Solid, 10 mg
Katalog # 786-473
Leverandør:   G-Biosciences
Jacalin for IgA purification
Katalog # 786-473
Leverandør:  G-Biosciences
CAS nummer:  
Jacalin for IgA purification
Jacalin for IgA purification
Jacalin, immobilised, lyophilised, Solid, 10 mg
CAS nummer:  
Price
1.540,00 kr
Log ind for tilgængelighed
1 * 10 mg
Product Details & Documents
Jacalin, or Artocarpus integrifolia lectin, is a tetrameric two-chain lectin with a molecular weight of 66 kDa. It is a α-D-galactose binding lectin purified from jack fruit (Artocarpus integrifolia) seeds.
Applications include isolating IgA from human serum and colostrums, isolating human plasma glycoproteins and histochemistry. It also binds IgD.
Specifikationer
Specifikationer
  • Pakn:
    10 mg
  • Beskrivelse:
    Jacalin, immobilised, lyophilised
  • Model:
    Solid
Specifikationer