Ammoniumchlorid
Plastic bottle for solids, 500 g
Katalog # RC-015
Leverandør: G-Biosciences
Gå til:
Product Details & Documents
Synonymer: Ammonium muriate, Salmiac
- CAS nummer:12125-02-9
- Formel:NH₄Cl
- MDL nummer:MFCD00011420
- Molecular Weight:53.49 g/mol
- Merck Index:13,00510
- Smeltende Pt:335 °C
- Tæthed:1,5256 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:235-186-4
Specifikationer
Specifikationer
- Pakning:Plastic bottle for solids
- Pakn:500 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



