Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
L-Alanin

L-Alanin

Glass bottle for solids, 250 g
Catalog # RC-014
Supplier:   G-Biosciences
L-Alanin
Catalog # RC-014
Supplier:  G-Biosciences
CAS Number:  56-41-7
L-Alanin

Glass bottle for solids, 250 g

CAS Number:  56-41-7
Price
1.630,00 kr
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
  • CAS Number:56-41-7
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00064410
  • Molecular Weight:89.09 g/mol
  • Merck Index:13,00203
  • Boiling Pt:189 °C (1013 hPa)
  • Melting Pt:79 °C
  • EINECS:206-418-1
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifications