Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ampicillin, natriumsalt

Ampicillin, natriumsalt

Glass bottle for solids, 25 g
Katalog # RC-020
Sundhed
Leverandør:   G-Biosciences
Ampicillin, natriumsalt
Katalog # RC-020
Leverandør:  G-Biosciences
CAS nummer:  69-52-3
Ampicillin, natriumsalt

Glass bottle for solids, 25 g

CAS nummer:  69-52-3
Price
1.500,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt
  • CAS nummer:69-52-3
  • Formel:C₁₆H₁₈N₃NaO₄S
  • MDL nummer:MFCD00064313
  • Molecular Weight:371.39 g/mol
  • Opbevaringstemperatur:Køleskab
  • EINECS:200-708-1
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.