Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

L-Alanin

Glass bottle for solids, 100 g
Katalog # RC-013
Leverandør:   G-Biosciences
L-Alanin
Katalog # RC-013
Leverandør:  G-Biosciences
CAS nummer:  56-41-7
L-Alanin

Glass bottle for solids, 100 g

CAS nummer:  56-41-7
Price
730,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  (S)-(+)-Alanine, H-Ala-OH, L(+)-Alanine, Alanine, L-α-Alanine, A, H-L-Ala-OH, α-Alanine
  • CAS nummer:56-41-7
  • Formel:C₃H₇NO₂
  • MDL nummer:MFCD00064410
  • Molecular Weight:89.09 g/mol
  • Merck Index:13,00203
  • Kogende Pt:189 °C (1013 hPa)
  • Smeltende Pt:79 °C
  • EINECS:206-418-1
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.