Orders LinkContactUsLinkComponent
Carbohydrate coupling resin
Carbohydrate coupling resin
Carbohydrate Immobilisation Kit, for 5×2 ml columns, 1 KIT
Catalog # 786-807
Supplier:   G-Biosciences
Carbohydrate coupling resin
Catalog # 786-807
Supplier:  G-Biosciences
CAS Number:  
Carbohydrate coupling resin
Carbohydrate coupling resin
Carbohydrate Immobilisation Kit, for 5×2 ml columns, 1 KIT
CAS Number:  
Price
5.690,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
Carbohydrate coupling resin is designed for the covalent immobilisation of carbohydrate containing molecules, including glycoproteins, to agarose beads. It is ideal for the coupling of polyclonal antibodies as it allows for the optimal orientation of the antibodies for affinity purification.
The Carbohydrate Immobilization Kit is supplied with carbohydrate coupling resin columns for the preparation of 2 ml affinity columns, a coupling catalyst, SpinOUT™ GT-600 Desalting Columns for removing oxidising reagents, the oxidizing reagent (sodium meta-periodate) and relevant buffers.
Specifications
Specifications
  • Pakn:
    1 KIT
  • Beskrivelse:
    Carbohydrate Immobilisation Kit, for 5×2 ml columns