Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Carboxyl coupling resin

Carboxyl coupling resin

Carboxyl Immobilisation Kit, for 5×2 ml columns, 1 KIT
  786-809
Carboxyl coupling resin
  786-809
 :  
Carboxyl coupling resin
Carboxyl coupling resin

Carboxyl Immobilisation Kit, for 5×2 ml columns, 1 KIT

 :  
 
4.940,00 kr
   
1 * 1 Each
 

Carboxyl coupling resin consists of 6% cross-linked agarose with covalently linked diaminodipropylamine (DADPA) to generate a free primary amine at the end of a long spacer arm. Molecules, including proteins, peptides and nucleic acids, are covalently coupled to the free primary amines, and the stable columns are ideal for affinity purification of antibodies and other interacting partners.

 

 

  • Pakn:
    1 KIT
  • Beskrivelse:
    Carboxyl Immobilisation Kit, for 5×2 ml columns

 

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.